Learn More
HLA-DR, DP, DQ Mouse anti-Human, BUV496, Clone: Tu39 (also known as TÜ39), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | HLA-DR, DP, DQ |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Tu39 (also known as TÜ39) |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Tü39; MHC class II HLA-DR, DP, DQ |
| Host Species | Mouse |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.