Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Mouse anti-Human, Alexa Fluor 488, Clone: L203.rMAb (also known as L203), BD Pharmingen™
Mouse Monoclonal Antibody
$132.00 - $285.50
Specifications
Antigen | HLA-DR |
---|---|
Clone | L203.rMAb (also known as L203) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow CytometrySpecifications
HLA-DR | |
Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
HLA-DR alpha beta; HLA-DR alpha,beta; HLA-DR alpha-beta; HLA-DR alpha/beta; HLA-DRαβ | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
L203.rMAb (also known as L203) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
RPMI 8866 human lymphoblastoid cells | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title