Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma,Dog, BB700, Clone: G46-6, BD Horizon™
Mouse Monoclonal Antibody
$204.00 - $444.00
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | G46-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human,Baboon,Cynomolgus,Rhesus Ma,Dog samples. It is validated for Flow Cytometry.Specifications
| HLA-DR | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human, Baboon, Cynomolgus, Rhesus Macaque, Dog | |
| Primary |
| G46-6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title