Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Antibody (37.68), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065424R
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human, Bovine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| HLA-DR | |
| Monoclonal | |
| DyLight 550 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-DRA | |
| Splenocytes from ATL mice (1) | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Human, Bovine, Goat, Sheep | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 37.68 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| 0.125 mL | |
| Primary | |
| NB100-65424 recognizes a monomorphic epitope on ovine MHC class II DR molecules. The distribution of ovine DR molecules on T lymphocytes has been shown to vary with immune status and age. Initially T lymphocytes lack class II molecules, then T lymphocytes progressively express DR molecules. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction