Learn More
Description
Specifications
Specifications
| Antigen | HLA-DR |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | L243 |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | MHC class II antigen; HLA class II histocompatibility antigen |
| Host Species | Mouse |
| Immunogen | Human lymphoblastoid B-cell line RPMI 8866 |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.