Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DR Mouse anti-Human, BV605, Clone: L243, BD Horizon™
Mouse Monoclonal Antibody
$243.00 - $482.00
Specifications
Antigen | HLA-DR |
---|---|
Clone | L243 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 605 |
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Flow cytometrySpecifications
HLA-DR | |
Flow Cytometry | |
Brilliant Violet 605 | |
Mouse | |
Human | |
MHC class II antigen; HLA class II histocompatibility antigen | |
IgG2a κ | |
Affinity Chromatography |
L243 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human lymphoblastoid B-cell line RPMI 8866 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title