Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRA Antibody (169-1B5.2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247722MFV500
Description
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human, Bovine, Feline samples. It is validated for Flow CytometrySpecifications
HLA DRA | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine, Feline | |
Purified |
Flow Cytometry | |
169-1B5.2 | |
Flow Cytometry | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Human peripheral blood leukocytes (Uniprot: P01903) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
3122 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction