Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRA Antibody (19-26.1 (same as MB-26.1)), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2450440.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, SDS-Page, Immunofluorescence.Specifications
HLA DRA | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This MAb reacts with the HLA-DR antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36kDa alpha (heavy) chain and a 28kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4+ T cells. It is an excellent histiocytic marker in paraffin sections producing intense cytoplasmic staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts. | |
Store at 4C. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
19-26.1 (same as MB-26.1) | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
HLA-DRA | |
Raji Cells | |
0.2 mg | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
3122 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction