Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DRA Mouse anti-Human, Unconjugated, Clone: TAL 1B5, Abnova™

Catalog No. 89337722 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-722 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-722 Supplier Abnova Corporation Supplier No. MAB21229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

HLA-DRA Monoclonal antibody specifically detects HLA-DRA in Human samples. It is validated for IHC-P, Flow Cyt.

Specifications

Antigen HLA-DRA
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone TAL 1B5
Conjugate Unconjugated
Description major histocompatibility complex, class II, DR alpha
Gene HLA-DRA
Gene Alias HLA-DRA1
Host Species Mouse
Immunogen Monoclonal antibody raised against Bristol 8 cell line of human origin
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3122
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.