Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DRB1 Mouse anti-Human, Unconjugated, Clone: LN-3 + HLA-DRB/1067, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB21061
Description
HLA-DRB1 Monoclonal antibody specifically detects HLA-DRB1 in Human samples. It is validated for WB-Ce, IHC-P, IF, Flow Cyt.Specifications
HLA-DRB1 | |
Monoclonal | |
Unconjugated | |
HLA-DRB1 | |
Mouse | |
100 μg | |
3123 | |
Store at 2 to 8°C | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
LN-3 + HLA-DRB/1067 | |
major histocompatibility complex, class II, DR beta 1 | |
DRB1, FLJ76359, HLA-DR1B, HLA-DRB, HLA-DRB1*, SS1 | |
Activated human peripheral blood mononuclear cells (LN-3 and HLA-DRB/1067) | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction