Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DRB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 158621AP150UL

Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
HLA-DRB1 | |
Polyclonal | |
Unconjugated | |
HLA-DRB1 | |
DR4, DRB1, DW2.2/DR2.2, HLA DR1B, HLA DRB, HLA DRB1, HLA DRB1*, HLA DRB2, HLA-DRB1, MHC class II antigen DRB1*15, SS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3123 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04229 | |
HLA-DRB1 | |
HLA-DRB1 Fusion Protein Ag8405 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction