Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRB3 Mouse anti-Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel, Clone: Bra30, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30716820UG
Description
HLA DRB3 Monoclonal specifically detects HLA DRB3 in Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
HLA DRB3 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain | |
Human REH cells (non-T, non-B leukemia cell line) | |
20 μg | |
Asthma, Immunology | |
3125 | |
Store at 4°C. | |
IgG2a κ |
Flow Cytometry, Immunofluorescence | |
Bra30 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine, Chimpanzee, Monkey, Baboon, Squirrel | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction