Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DRB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598351
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598351 Supplier Invitrogen™ Supplier No. PA598351
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HLA-DRB3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Major histocompatibility complex (MHC) class II molecules destined for presentation to CD4+ helper T cells is determined by two key events. These events include the dissociation of class II-associated invariant chain peptides (CLIP) from an antigen binding groove in mhc ii-a/b dimers through the activity of MHC molecules HLA-DM and -DO, and subsequent peptide antigen binding. Accumulating in endosomal/lysosomal compartments and on the surface of B cells, HLA-DM, -DO molecules regulate the dissociation of CLIP and the subsequent binding of exogenous peptides to HLA class II molecules (HLA-DR, -DQ and -DP) by sustaining a conformation that favors peptide exchange. RFLP analysis of HLA-DM genes from rheumatoid arthritis (RA) patients suggests that certain polymorphisms are genetic factors for RA susceptibility. HLA-B belongs to the HLA class I heavy chain paralogs. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. HLA-B and -C can form heterodimers consisting of a membrane anchored heavy chain and a light chain ( beta-2-Microglobulin). Polymorphisms yield hundreds of HLA-B and -C alleles. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DRB3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HLA-DRB3
Gene Accession No. P79483
Gene Alias DR7; DRB1; DRw10; HLA class II histocompatibility antigen, DR beta 3 chain; HLA class II histocompatibility antigen, DRB1-7 beta chain; HLA DR3B; HLA-DR1B; HLA-DR3B; HLA-DRB; HLA-DRB3; human leucocyte antigen DRB3; major histocompatibility complex, class II, DR beta 3; MGC117330; MHC Class II antigen; MHC class II antigen DR beta 3 chain; MHC class II antigen DRB3; MHC class II HLA-DR beta 3 chain; SS1
Gene Symbols HLA-DRB3
Host Species Rabbit
Immunogen Recombinant Human HLA class II histocompatibility antigen, DR beta 3 chain protein (111-227AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3125
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.