Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | HLA DRB3 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HLA DRB3 Polyclonal specifically detects HLA DRB3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HLA DRB3 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DR7, HLA class II histocompatibility antigen, DR beta 3 chain, HLA class II histocompatibility antigen, DRB1-7 beta chain, HLA-DR3B, HLA-DR52, HLA-DRB1, human leucocyte antigen DRB3, major histocompatibility complex, class II, DR beta 3, MGC117330, MHC class II antigen DR beta 3 chain, MHC class II antigen DRB3, MHC class II HLA-DR beta 3 chain | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-227 of human HLA DRB3 (NP_072049.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Asthma, Immunology | |
PBS with 50% glycerol, pH7.3. | |
3125 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title