Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRB4 Rabbit anti-Human, Clone: 3Z5V9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | HLA DRB4 |
---|---|
Clone | 3Z5V9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
HLA DRB4 Monoclonal antibody specifically detects HLA DRB4 in Human samples. It is validated for Western BlotSpecifications
HLA DRB4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Asthma, Immunology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3126 | |
IgG | |
Affinity purified |
3Z5V9 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
class II histocompatibility antigen HLA DR alpha, beta1-0307, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR4, DR-4, DRB1 transplantation antigen, DRB4, HLA class II histocompatibility antigen, DR beta 4 chain, HLA class II histocompatibility antigen, DRB1-12 beta chain, HLA class II histocompatibility antigen, DRB1-4 beta chain, HLA-DR4B, human leucocyte antigen DRB4, leucocyte antigen DRB1, leukocyte antigen, major histocompatibility complex, class II, DR beta 1, major histocompatibility complex, class II, DR beta 4, MHC class I antigen DRB1*12, MHC class I antigen DRB1*4, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*4, MHC class II antigen DRB4, MHC class II antigen HLA-DRB1, MHC class II antigen HLA-DR-beta, MHC class II HLA-DR-beta-7, MHC class2 antigen, MHC HLA DR-beta chain | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HLA DRB4 (P13762). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title