Learn More
Invitrogen™ HLA-DRB4 Recombinant Rabbit Monoclonal Antibody (3Z5V9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542594
Description
Positive test controls include: Raji. The target is usually found in the following locations: Cell membrane, Endoplasmic reticulum membrane, Endosome membrane, Golgi apparatus, Late endosome membrane, Lysosome membrane, Single-pass type I membrane protein, trans-Golgi network membrane. Immunogen sequence: TERVWNLIRY IYNQEEYARY NSDLGEYQAV TELGRPDAEY WNSQKDLLER RRAEVDTYCR YNYGVVESFT VQRRVQPKVT VYPSKTQPLQ HHNLLVCSVN G.
HLA-DRB4 belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DRA) and a beta (DRB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. DRB1 is expressed at a level five times higher than its paralogues DRB3, DRB4 and DRB5. The presence of DRB4 is linked with allelic variants of DRB1, otherwise it is omitted. There are 4 related pseudogenes: DRB2, DRB6, DRB7, DRB8 and DRB9. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
HLA-DRB4 | |
Recombinant Monoclonal | |
0.4 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13762 | |
HLA-DRB4 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human HLA-DRB4 (P13762). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
3Z5V9 | |
Unconjugated | |
HLA-DRB4 | |
DR4; DR-4; DRB1 transplantation antigen; DRB4; HLA class II histocompatibility antigen, DR beta 4 chain; HLA-DR4B; HLA-DRB4; human leucocyte antigen DRB4; leukocyte antigen; major histocompatibility complex, class II, DR beta 4; MHC class II antigen DRB4; MHC class II antigen HLA-DR-beta; MHC class2 antigen; MHC HLA DR-beta chain | |
Rabbit | |
Affinity chromatography | |
RUO | |
3126 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.