Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRB5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | HLA DRB5 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HLA DRB5 Polyclonal specifically detects HLA DRB5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HLA DRB5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
DR beta-5, DR2-beta-2, DR9, DR-9, dw2, FLJ76359, HLA class II histocompatibility antigen, DR beta 5 chain, HLA class II histocompatibility antigen, DR-5 beta chain, HLA class II histocompatibility antigen, DRB1-9 beta chain, HLA-DRB, human leucocyte antigen DRB5, leukocyte antigen class II, major histocompatibility complex, class II, DR beta 5, MHC class II antigen DRB1*9, MHC class II antigen DRB5, MHC class II HLA beta chain, MHC HLA-DR-beta cell surface glycoprotein, MHC HLA-DR-beta chain | |
HLA-DRB5 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Asthma, Diabetes Research, Immunology | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
3127 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GVGESFTVQRRVEPKVTVYPARTQT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title