Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DRB5 Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812174


Description
HLA-DRB5 Monoclonal antibody specifically detects HLA-DRB5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
HLA-DRB5 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5FVL0, Q80SY3, Q8N8Y2 | |
Mouse | |
Protein A/G | |
RUO | |
242341, 245972, 297932 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4G7 | |
Unconjugated | |
ATP6V0D2 | |
DR beta-5; DR2-beta-2; DR9; DR-9; DRB1-9 beta chain; dw2; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; hla-dr5; HLA-DRB; HLA-DRB5; human leucocyte antigen DRB5; leukocyte antigen class II; major histocompatibility complex, class II, DR beta 5; MHC class I antigen; MHC class II antigen DRB1*9; MHC class II antigen DRB5; MHC class II HLA beta chain; MHC HLA-DR; MHC HLA-DR-beta cell surface glycoprotein; MHC HLA-DR-beta chain | |
Full length human recombinant protein of human ATP6V0D2 (NP_689778) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction