Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DRB5 Polyclonal Antibody
HLA-DRB5 Polyclonal Antibody
Supplier: Thermo Scientific PA560260
Description
HLA-DRB5 Polyclonal Antibody for Western Blot, IHC

Specifications
HLA-DRB5 | |
Polyclonal | |
Unconjugated | |
HLA-DRB5 | |
DR beta-5; DR2-beta-2; DR9; DR-9; DRB1-9 beta chain; dw2; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; hla-dr5; HLA-DRB; HLA-DRB5; human leucocyte antigen DRB5; leukocyte antigen class II; major histocompatibility complex, class II, DR beta 5; MHC class I antigen; MHC class II antigen DRB1*9; MHC class II antigen DRB5; MHC class II HLA beta chain; MHC HLA-DR; MHC HLA-DR-beta cell surface glycoprotein; MHC HLA-DR-beta chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3127 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q30154 | |
HLA-DRB5 | |
Recombinant protein corresponding to Human HLA-DRB5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction