Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DRB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578004 Supplier Invitrogen™ Supplier No. PA578004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Raji, Raji membrane fraction extract Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HLA-DRB5 belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DRA) and a beta (DRB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. DRB1 is expressed at a level five times higher than its paralogues DRB3, DRB4 and DRB5. The presence of DRB5 is linked with allelic variants of DRB1, otherwise it is omitted. There are 4 related pseudogenes: DRB2, DRB6, DRB7, DRB8 and DRB9. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DRB5
Applications Western Blot
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene HLA-DRB5
Gene Accession No. Q30154
Gene Alias DR beta-5; DR2-beta-2; DR9; DR-9; DRB1-9 beta chain; dw2; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; hla-dr5; HLA-DRB; HLA-DRB5; human leucocyte antigen DRB5; leukocyte antigen class II; major histocompatibility complex, class II, DR beta 5; MHC class I antigen; MHC class II antigen DRB1*9; MHC class II antigen DRB5; MHC class II HLA beta chain; MHC HLA-DR; MHC HLA-DR-beta cell surface glycoprotein; MHC HLA-DR-beta chain
Gene Symbols HLA-DRB5
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human HLA-DRB5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3127
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.