Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DRw52 Monoclonal Antibody (7.3.19.1)
GREENER_CHOICE

Catalog No. 01671477
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-477 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-477 Supplier Invitrogen™ Supplier No. MA135422
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody reacts with a polymorphic determinant on HLA-DR molecules which is associated with DR3, DR5, and DRw6 (DRw52-like). It does not react with DRw8. The antibody reacts with part of the HLA-D molecules (29+34 kD). The determinant is strongly expressed on DR3 positive cells and more weakly on DR5 and DRw6 positive cells. The antibody is useful for HLA-DR typing and biochemical characterization of the DR52w positive molecules and functional studies. Cross reacts with Chimpanzee, Macaca-apes and gorilla.

HLA DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an alpha chain (36 kDa) and a beta chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DRw52
Applications Cytotoxicity Assay, Immunocytochemistry
Classification Monoclonal
Clone 7.3.19.1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene HLA-DRB1
Gene Accession No. P01911
Gene Alias Clone P2-beta-3; DR-16; DRB1; DRw10; DW2.2/DR2.2; FLJ75017; FLJ76359; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; lymphocyte antigen DRB1; major histocompatibility complex, class II, DR beta 1; MHC class II antigen; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; SS1; XXbac-BPG161M6.1
Gene Symbols HLA-DRB1
Host Species Mouse
Immunogen Human HLA-DRw52.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3123
Target Species Human, Non-human Primate
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.