Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-E Antibody (MEM-E/07), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500310PECY7
Description
HLA-E Monoclonal specifically detects HLA-E in Human samples. It is validated for Flow Cytometry.Specifications
HLA-E | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
HLA-E | |
Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide. | |
0.1 mL | |
Primary | |
The antibody MEM-E/07 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that the antibody cross-reacts with some classical MHC Class I molecules (MHC Class Ia): HLA-B7 (strongly), HLA-B8 (moderately), HLA-B27, -B44 (weakly). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MEM-E/07 | |
Flow Cytometry | |
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Asthma, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
3133.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction