Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-E Mouse anti-Human, R718, Clone: 3D12 (also known as 3D12/HLA-E), BD Horizon™
Mouse Monoclonal Antibody
$246.00 - $452.00
Specifications
Antigen | HLA-E |
---|---|
Clone | 3D12 (also known as 3D12/HLA-E) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Flow CytometrySpecifications
HLA-E | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
HLA class I histocompatibility antigen, alpha chain E; HLAE; MHC Class I Antigen E | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
3D12 (also known as 3D12/HLA-E) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HLA-E Recombinant Protein | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title