Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA F Antibody (3D11), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP276802APCCY7
Description
HLA F Monoclonal specifically detects HLA F in Human samples. It is validated for Flow Cytometry.Specifications
| HLA F | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% sodium azide | |
| HLA-F | |
| Inclusion body-derived HLA-F heavy chain | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 3D11 | |
| Flow Cytometry | |
| alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen | |
| Mouse | |
| 0.1 mL | |
| 3134 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction