Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA F Antibody (SR1707), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HLA F |
---|---|
Clone | SR1707 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
HLA F Monoclonal antibody specifically detects HLA F in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
HLA F | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen | |
A synthesized peptide derived from human HLA F (Uniprot #: P30511) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1707 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3134 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title