Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-F Monoclonal Antibody (2010CT409.22.10)
GREENER_CHOICE

Catalog No. PIMA537773
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537773 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537773 Supplier Invitrogen™ Supplier No. MA537773
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLA-F Monoclonal Antibody for Western Blot

HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-F
Applications Western Blot
Classification Monoclonal
Clone 2010CT409.22.10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HLA-F
Gene Accession No. P30511
Gene Alias CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen
Gene Symbols HLA-F
Host Species Mouse
Immunogen recombinant protein from the human region of human HLA-F.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.