Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-F, Mouse anti-Human, Clone: 3D11, Millipore Sigma™

Catalog No. MABF216225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF216225 25 μg
MABF2162100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF216225 Supplier MilliporeSigma Supplier No. MABF216225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HLA-F, clone 3D11, Cat. No. MABF2162, is a mouse monoclonal antibody that detects HLA class I histocompatibility antigen, alpha chain F and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

HLA class I histocompatibility antigen, alpha chain F (UniProt: P30511; also known as CDA12, HLA F antigen, Leukocyte antigen F, MHC class I antigen F) is encoded by the HLA-F (also known as HLA-5.4, HLAF) gene (Gene ID: 3134) in human. HLA-F is a single-pass type I membrane heterodimeric protein that is involved in the presentation of foreign antigens to the immune system. It is expressed in B cells, NK cells, monocytes, and T cells and its expression is reported to be coincident with the activated immune response. It is mainly localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off. The mature form has an extracellular domain (aa 22-305), a transmembrane domain (aa 306-329), and a short cytoplasmic domain (aa 330-346). The HLA-F locus has very low allelic polymorphism and is highly conserved in distantly related non-human primates indicating its conserved function. It is involved in the regulation of immune system in pregnancy, infection, and autoimmunity by signaling through NK cell receptors. (Ref.: Dulberger, CL et al. (2017). Immunity 46(6); 1018 1029; Lee, N., et al. (2010). Eur. J. Immunol. 40(8): 2308 2318).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-F
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3D11
Conjugate Unconjugated
Dilution ELISA Analysis: A representative lot detected HLA-F in ELISA applications (Lee, N., et. al. (2010). Eur J Immunol. 40(8):2308-18).Western Blotting Analysis: A representative lot detected HLA-F in Western Blotting applications (Lee, N., et. al. (2010). Eur J Immunol. 40(8):2308-18; Ishitani, A., et. al. (2003). J Immunol. 171(3):1376-84).Flow Cytometry Analysis: A representative lot detected HLA-F in Flow Cytometry applications (Lee, N., et. al. (2010). Eur J Immunol. 40(8):2308-18).Immunohistochemistry Analysis: A representative lot detected HLA-F in Immunohistochemistry applications (Ishitani, A., et. al. (2003). J Immunol. 171(3):1376-84).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias HLA class I histocompatibility antigen alpha chain F;CDA12;HLA F antigen;Leukocyte antigen F;MHC class I antigen F
Gene Symbols HLA-F;HLA-5.4;HLAF
Host Species Mouse
Immunogen Full-length human recombinant HLA-F Heavy chains (excluding the signal sequence).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_061823
Test Specificity Clone 3D11 specifically detects human HLA-F. It targets an epitope with in the alpha 2 domain from the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.