Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587858 Supplier Invitrogen™ Supplier No. PA587858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSHSLRYFST AVSRPGRGEP RYIAVEYVDD TQFLRFDSDA AIPRMEPREP WVEQEGPQYW EWTTGYAKAN AQTDRVALRN LLRRYNQSEA GSHTLQGMNG CDMGPDGRLL RGYHQHAYDG KDYISLNEDL RSWTAADTVA QITQRFYEAE EYAEEFRTYL EGECLELLR; Positive Samples: A-549; Cellular Location: Membrane, Single-pass type I membrane protein.

HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-F
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HLA-F
Gene Accession No. P30511
Gene Alias CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen
Gene Symbols HLA-F
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-190 of human HLA-F (NP_0010919491).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3134
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.