Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA G Antibody (01G), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144926H
Description
HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
HLA G | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
HLA-G | |
HLA-B27 transgenic mice were imunized with H-2 identical murine cells transfected with and expressing genes encoding HLA-G and human beta2-microglobulin. | |
0.1 mL | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
The antibody 01G recognizes membrane-bound form of HLA-G (full-length HLA-G1), but not soluble forms. HLA-G belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on the surface of trophoblast cells. | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
01G | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction