Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA G Antibody (17A5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | HLA G |
---|---|
Clone | 17A5 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Flow Cytometry, ELISASpecifications
HLA G | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G | |
A synthesized peptide derived from Human HLA G [UniProt P17693] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
17A5 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3135 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title