Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA G Antibody (87G), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143123APCCY7
Description
HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Flow Cytometry.Specifications
HLA G | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
HLA-G | |
721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt No. P17693] | |
0.1 mL | |
Primary | |
Membrane bound HLA G1 and soluble HLA G5. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
87G | |
Flow Cytometry | |
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G | |
Mouse | |
Protein G purified | |
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
3135.0 | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction