Learn More
HLA G Antibody (87G), DyLight 550, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP143123R
Catalog No. NBP143123R
Description
Description
HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HLA G
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
HLA-G
721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt# P17693]
0.1 mL
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
3135
Human, Mouse
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
87G
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G
Mouse
Protein G purified
RUO
Primary
Membrane bound HLA G1 and soluble HLA G5.
Store at 4C in the dark.
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only