Learn More
Invitrogen™ HLA-G Monoclonal Antibody (G233)

Description
This antibody recognizes an extracellular epitope on several isoforms of HLA-G expressed in all populations of extravillous trophoblast (cell columns, interstitial trophoblast, endovascular trophoblast, placental bed giant cells). HLA-G belongs to the nonclassical MHC Class I molecules (MHC Class Ib). The antibody G233 has been found not to cross-react with any other MHC Class I antigens (HLA-A, -B, -C, -E, -F).
Specifications
Specifications
| Antigen | HLA-G |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | G233 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | HLA-G |
| Gene Accession No. | P17693 |
| Gene Alias | acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.