Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLAA (HLA-A) Monoclonal Antibody (OTI3H6), TrueMAB™, OriGene
SDP

Catalog No. 502543668
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3668 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3668 Supplier OriGene Supplier No. CF813379
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HLAA (HLA-A) Monoclonal antibody specifically detects HLAA (HLA-A) in Human samples. It is validated for Western Blot

Specifications

Antigen HLAA (HLA-A)
Applications Western Blot
Classification Monoclonal
Clone OTI3H6
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HLA-A
Gene Accession No. P04439
Gene Alias DR beta-5; DR2-beta-2; DR9; DR-9; DRB1-9 beta chain; dw2; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; hla-dr5; HLA-DRB; HLA-DRB5; human leucocyte antigen DRB5; leukocyte antigen class II; major histocompatibility complex, class II, DR beta 5; MHC class I antigen; MHC class II antigen DRB1*9; MHC class II antigen DRB5; MHC class II HLA beta chain; MHC HLA-DR; MHC HLA-DR-beta cell surface glycoprotein; MHC HLA-DR-beta chain
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 25-308 of human HLA-A (NP_002107) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.