Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLC8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9386 Supplier Bioss Supplier No. BS9836R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HLC8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C17orf80
Gene Accession No. Q9BSJ5
Gene Alias C17orf80; Cell migration-inducing gene 3 protein; chromosome 17 open reading frame 80; HLC8; HLC-8; Human lung cancer oncogene 8 protein; lung cancer-related protein 8; MIG3; SPEP1; sperm-expressed protein 1; Uncharacterized protein C17orf80
Gene Symbols C17orf80
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HLC8/Lung cancer related protein 8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55028
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.