Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLCS Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299591FR
Description
HLCS Polyclonal specifically detects HLCS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HLCS | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Biotin apo-protein ligase, biotin--protein ligase, EC 6.3.4.-, HCSholocarboxylase synthetase, EC 6.3.410holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carboxylase(ATP-hydrolysing)) ligase), holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase(ATP-hydrolysing)) ligase), holocarboxylase synthetase (biotin-[proprionyl-Coenzyme A-carboxylase(ATP-hydrolysing)] ligase) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HLCS. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
3141 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction