Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLCS Antibody (OTI1E4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270893PCP
Description
HLCS Monoclonal specifically detects HLCS in Human samples. It is validated for Flow Cytometry.Specifications
| HLCS | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| HLCS | |
| Full length recombinant protein of human HLCS (NP_000402) produced in HEK293T cells. | |
| 0.1 mL | |
| 3141.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1E4 | |
| Flow Cytometry | |
| Biotin apo-protein ligase, biotin--protein ligase, EC 6.3.4.-, HCSholocarboxylase synthetase, EC 6.3.410holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carboxylase(ATP-hydrolysing)) ligase), holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase(ATP-hydrolysing)) ligase), holocarboxylase synthetase (biotin-[proprionyl-Coenzyme A-carboxylase(ATP-hydrolysing)] ligase) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction