Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLCS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553599 Supplier Invitrogen™ Supplier No. PA553599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLPYDYSSSL ESVADETSPE REGRRVNLTG KAPNILLYVG SDSQEALGRF HEVRSVLADC VDIDSYILYH LLEDSALRDP WTDNCLLLVI ATRESIPEDL YQKFMAYLSQ GGKVLGLSSS FTFGGFQV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.

The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae Cdc23, a protein essential for cell cycle progression through the G2/M transition. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLCS
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HLCS
Gene Accession No. P50747
Gene Alias 410I21.SP6; biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase; Biotin apo-protein ligase; biotin--[acetyl-CoA-carboxylase] ligase; Biotin--[methylcrotonoyl-CoA-carboxylase] ligase; biotin--[methylmalonyl-CoA-carboxytransferase] ligase; Biotin--[propionyl-CoA-carboxylase [ATP-hydrolyzing]] ligase; Biotin--protein ligase; D16Jhu34; HCS; Hlcs; Holocarboxylase synthetase; holocarboxylase synthetase (biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase); holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase); holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carboxylase (ATP-hydrolysing)) ligase); RGD1565360
Gene Symbols HLCS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HLCS. Recombinant protein control fragment (Product #RP-91775).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.