Learn More
Invitrogen™ HLF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595523
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, rat brain tissue, rat liver tissue, rat kidney tissue, rat lung whole cell, mouse brain tissue, mouse liver tissue, mouse kidney tissue, mouse lung tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the proline and acidic-rich (PAR) protein family, a subset of the bZIP transcription factors. The encoded protein forms homodimers or heterodimers with other PAR family members and binds sequence-specific promoter elements to activate transcription. Chromosomal translocations fusing portions of this gene with the E2A gene cause a subset of childhood B-lineage acute lymphoid leukemias. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
HLF | |
Polyclonal | |
Unconjugated | |
HLF | |
E230015K02Rik; hepatic leukemia factor; HLF; HLF transcription factor, PAR bZIP family member; HLF, PAR bZIP transcription factor; LOC690286; PAR bZIP transcription factor; similar to hepatic leukemia factor | |
Rabbit | |
Affinity chromatography | |
RUO | |
217082, 3131, 690286 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
Q16534, Q64709, Q8BW74 | |
HLF | |
A synthetic peptide corresponding to a sequence of human HLF (DDLKDDKYWARRRKNNMAAKRSRDARRLKE). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.