Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ HLI 373
GREENER_CHOICESDP

Catalog No. 350350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-035-0 50 mg
35-031-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-035-0 Supplier R&D Systems™ Supplier No. 3503/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Hdm2 inhibitor; activates p53-dependent transcription

Inhibitor of Hdm2 ubiquitin ligase (E3). Blocks Hdm2-mediated ubiquitylation and proteasomal degradation of p53; activates p53-dependent transcription. Induces apoptosis in several tumor cell lines that express wild-type p53 such as LOX-IMVI, A549, HT1080 and U2OS.

Chemical Identifiers

CAS 1782531-99-0

Specifications

Quantity 50 mg
Product Line Tocris
Molecular Formula C18H23N5O2
Formula Weight 414.33
Purity >99%
Molecular Weight (g/mol) 341.415
Chemical Name or Material HLI 373
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.