Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLTF Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100280AF350
Description
HLTF Polyclonal antibody specifically detects HLTF in Human samples. It is validated for Western BlotSpecifications
HLTF | |
Polyclonal | |
Western Blot | |
DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
A synthetic peptide, which represented a portion of human SMARCA3 encoded in part by exons 23 and 24 (LocusLink ID 6596). | |
0.1 mL | |
Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
6596 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction