Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLTF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100280
Description
HLTF Polyclonal specifically detects HLTF in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HLTF | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
HLTF | |
A synthetic peptide, which represented a portion of human SMARCA3 encoded in part by exons 23 and 24 (LocusLink ID 6596). | |
100 μL | |
Zinc Finger | |
6596 | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 -10 ug/mg of lysate | |
DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for human SMARCA3. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction