Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLTF Rabbit anti-Human, Mouse, Rat, Clone: 8T0I2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | HLTF |
---|---|
Clone | 8T0I2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
HLTF Monoclonal antibody specifically detects HLTF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HLTF | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Chromatin Research, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6596 | |
IgG | |
Affinity purified |
8T0I2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DNA-binding protein/plasminogen activator inhibitor 1 regulator, EC 3.6.1, EC 3.6.4.-, EC 6.3.2.-, helicase-like transcription factor, HIP116ARING finger protein 80, HIP116matrix associated, actin dependent regulator of chromatin, RNF80SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 3, subfamily a, member 3, Sucrose nonfermenting protein 2-like 3, ZBU1 | |
A synthetic peptide corresponding to a sequence within amino acids 900-1009 of human HLTF (Q14527). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title