Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596674 Supplier Invitrogen™ Supplier No. PA596674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HPHASFQAAA RSPLRPTPVV APSEVPAGFP QRLSPLSAAY HHHHPQQQQQ QQQPQQQQPP PPPRAGALQP PASGTRVVPN PHHSGSAPAP SSKDLKFGID RILSAEFDPK VKEGNTLRDL TSLLTGGRPA GVHLSGLQPS AGQFFASLDP INEASAILSP LNSNPRNSVQ HQFQDTFPGP YAVLTKDTMP QTYKRKRSWS R; Positive Samples: HL-60, Raji, THP-1, 293T; Cellular Location: Nucleus.

HLX is a transcription factor required for TBX21/T-bet-dependent maturation of Th1 cells as well as maintenance of Th1-specific gene expression. Involved in embryogenesis and hematopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLX
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HLX
Gene Accession No. Q14774
Gene Alias dbx1a; developing brain homeobox 1a; developing brain homeobox protein 1-A; fc15c04; H2.0 homeobox 1; H2.0 like homeobox; H2.0-like homeo box 1; H2.0-like homeo box-1; H2.0-like homeobox; H2.0-like homeobox 1; H2.0-like homeobox protein; HB24; Hlx; hlx1; hlx-1; homeobox protein; homeobox protein DBX1-A; Homeobox protein HB24; homeobox protein HLX1; LOW QUALITY PROTEIN: H2.0-like homeobox protein; wu:fc15c04; zgc:109861
Gene Symbols HLX
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-280 of human HLX (NP_0687771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3142
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.