Learn More
HLXB9 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303185A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Homeobox protein HB9 is expressed in lymphoid and pancreatic tissues. It is reported to be Putative transcription factor involved in pancreas development and function. Defects in this gene causes Currarino syndrome. The triad of a presacral tumor, sacral agenesis and anorectal malformation constitutes the Currarino syndrome which is caused by dorsal-ventral patterning defects during embryonic development. It is also reported that HB9 is strongly expressed in epidermal basal cells of the outer scale face in tarsometatarsal scale skin.Specifications
| HLXB9 | |
| Polyclonal | |
| Unconjugated | |
| MNX1 | |
| HB9, HLXB9, HOXHB9, SCRA1, homeobox HB9, homeobox protein HB9, motor neuron and pancreas homeobox protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3110 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P50219 | |
| MNX1 | |
| Between 351 and 401 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.