Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hLYVE-1, Mouse anti-Human, Clone: 8C, Millipore Sigma™

Catalog No. MABS192925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS192925 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS192925 Supplier MilliporeSigma Supplier No. MABS192925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-hLYVE-1, clone 8C, Cat. No. MABS1929, is a mouse monoclonal antibody that detects Lymphatic vessel endothelial hyaluronic acid receptor 1 (LYVE1) and has been tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.

Lymphatic vessel endothelial hyaluronic acid receptor 1 (UniProt: Q9Y5Y7; LYVE-1, Cell surface retention sequence-binding protein 1, CRSBP-1,Extracellular link domain-containing protein 1,Hyaluronic acid receptor) is encoded by the LYVE1 (also known as CRSBP1, HAR, XLKD1,UNQ230/PRO263) gene (Gene ID: 10894) in human. LYVE-1 is a disulfide-linked, homodimeric, single-pass type I membrane protein that undergoes ligand-dependent internalization and recycling at the cell surface.  It serves as a ligand-specific transporter trafficking between intracellular organelles (TGN) and the plasma membrane.  It is mainly expressed in endothelial cells lining lymphatic vessels. It is expressed on both the luminal and basolateral surfaces of lymphatic endothelium. It is reported to mediate the uptake of intermediate and high molecular weight hyaluronan (HA) and serves as a low affinity receptor for HA-induced lymphoangiogenesis. It also controls the entry and/or migration of leukocytes in lymphatic vessels. The capacity of LYVE-1 to bind HA is based on a receptor distribution and organization of the glycosaminoglycans. (Ref.: Lawrence, W et al. (2016). J Biol Chem. 291(15): 8014 8030).
TRUSTED_SUSTAINABILITY

Specifications

Antigen hLYVE-1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8C
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected hLYVE-1 in Immunocytochemistry applications (Lawrance, W., et. al. (2016). J Biol Chem. hLYVE-1; Banerji, S., et. al. (1999). J Cell Biol. 144(4):789-801). Western Blotting Analysis: A representative lot detected hLYVE-1 in Western Blotting applications (Banerji, S., et. al. (1999). J Cell Biol. 144(4):789-801; Banerji, S., et. al. (2016). J Biol Chem. 291(48):25004-25018).Flow Cytometry Analysis: A representative lot detected hLYVE-1 in Flow Cytometry applications (Lawrance, W., et. al. (2016). J Biol Chem. hLYVE-1; Banerji, S., et. al. (2016). J Biol Chem. 291(48):25004-25018).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Lymphatic vessel endothelial hyaluronic acid receptor 1;Cell surface retention sequence-binding protein 1;CRSBP-1;Extracellular link domain-containing protein 1;Hyaluronic acid receptor
Gene Symbols LYVE1;CRSBP1;HAR;XLKD1;UNQ230/PRO263
Host Species Mouse
Immunogen A recombinant fragment corresponding to the ectodomain (1-232) region of human LYVE-1.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 8C detects Lymphatic vessel endothelial hyaluronic acid receptor 1. It targets an epitope within the ectodomain region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.