Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Catalog No. p-4529979 Shop All Epredia Products
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Standard Knife Holder
For Use With (Equipment):
HM 525 NX and HM 550
Catalog No. 14-070-38
Supplier: Epredia™ 705460
Only null left
Add to Cart
Add to Cart
Request a Quote
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Standard Knife Carrier:
  • Allows for insertion of standard knives, with clearance angle adjustment
  • Can be moved horizontally on precision dovetail guide with quick clamp system
  • Includes anti-roll plate
Disposable Blade Carrier EC:
  • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
  • Can be moved on precision dovetail guide with quick clamp plate
  • Includes anti-roll plate
Disposable Blade Carrier EV:
  • Use with vacutome system
  • Allows for insertion of high- and low-profile disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
  • Use with vacutome system
  • Allows for insertion of disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Product Type Standard Knife Holder
For Use With (Equipment) HM 525 NX and HM 550
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.