Learn More
Invitrogen™ HM13 Polyclonal Antibody

Description
Immunogen sequence: PASFPNRQYQ LLFTQGSGEN KEEIINYEFD TKDLVCLGLS SIVGVWYLLR KHWIANNLFG LAFSLNGVEL LHLNNVSTGC Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Specifications
Specifications
| Antigen | HM13 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HM13 |
| Gene Accession No. | Q8TCT9 |
| Gene Alias | 1200006O09Rik; 4930443L17Rik; 5031424B04Rik; AV020344; cb228; H13; H-13; hIMP1; histocompatibility (minor) 13; histocompatibility 13; histocompatibility minor 13; HM13; hm13 protein; IMP1; IMP-1; IMPAS; IMPAS-1; intramembrane aspartyl protease; intramembrane protease 1; minor histocompatibility antigen 13; minor histocompatibility antigen H13; MSTP086; presenilin-like aspartyl protease; presenilin-like protein 3; PSENL3; PSL3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1; SPP; SPPL1; Unknown (protein for MGC:134054); wu:fc11a06; zgc:56660; zgc:86862 |
| Gene Symbols | HM13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.