Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HM13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569652 Supplier Invitrogen™ Supplier No. PA569652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 92%.

Catalyzes intramembrane proteolysis of some signal peptides after they have been cleaved from a preprotein, resulting in the release of the fragment from the ER membrane into the cytoplasm. Required to generate lymphocyte cell surface (HLA-E) epitopes derived from MHC class I signal peptides. May play a role in graft rejection. May be necessary for the removal of the signal peptide that remains attached to the hepatitis C virus core protein after the initial proteolytic processing of the polyprotein. Involved in the intramembrane cleavage of the integral membrane protein PSEN1. Cleaves the integral membrane protein XBP1 isoform 1 in a DERL1/RNF139- dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HM13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene H13
Gene Accession No. Q9D8V0
Gene Alias 1200006O09Rik; 4930443L17Rik; 5031424B04Rik; AV020344; cb228; H13; H-13; hIMP1; histocompatibility (minor) 13; histocompatibility 13; histocompatibility minor 13; HM13; hm13 protein; IMP1; IMP-1; IMPAS; IMPAS-1; intramembrane aspartyl protease; intramembrane protease 1; minor histocompatibility antigen 13; minor histocompatibility antigen H13; MSTP086; presenilin-like aspartyl protease; presenilin-like protein 3; PSENL3; PSL3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1; SPP; SPPL1; Unknown (protein for MGC:134054); wu:fc11a06; zgc:56660; zgc:86862
Gene Symbols H13
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse HM13 (aa 240-289).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14950
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.