Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212358F
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.Specifications
HM74A/PUMA-G/GPR109A/NIACR1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HCAR2 | |
Human recombinant HM74A/PUMA-G/GPR109A/NIACR1 | |
0.1 mL | |
338442 | |
Store at 4C in the dark. | |
IgG2 |
Flow Cytometry, Immunohistochemistry | |
MM0329-6S24 | |
Flow Cytometry, Immunohistochemistry | |
GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 | |
Rat | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction